Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172320.1 | NP_758823.1 | 2 - 3 | 142 | 104 | |
XM_008770001.1 | XP_008768223.1 | 2 - 3 | 190 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |