Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012499.1 | NP_036631.1 | 4 - 5 | 577 | 80 | |
XM_006254506.2 | XP_006254568.1 | 4 - 5 | 783 | 80 | ||
XM_006254508.2 | XP_006254570.1 | 5 - 6 | 624 | 80 | ||
XM_006254509.2 | XP_006254571.1 | 5 - 6 | 593 | 80 | ||
XM_006254510.2 | XP_006254572.1 | 4 - 5 | 783 | 80 | ||
XM_006254511.2 | XP_006254573.1 | 5 - 6 | 624 | 80 | ||
XM_006254512.2 | XP_006254574.1 | 4 - 5 | 1232 | 80 | ||
XM_006254513.2 | XP_006254575.1 | 4 - 5 | 783 | 80 | ||
XM_006254515.2 | XP_006254577.1 | 5 - 6 | 449 | 80 | ||
XM_006254518.2 | XP_006254580.1 | 4 - 5 | 1047 | 80 | ||
XM_006254520.2 | XP_006254582.1 | 4 - 5 | 783 | 80 | ||
XM_006254521.2 | XP_006254583.1 | 5 - 6 | 624 | 80 | ||
XM_008772015.1 | XP_008770237.1 | 4 - 5 | 2715 | 80 | ||
XM_008772016.1 | XP_008770238.1 | 4 - 5 | 2665 | 80 | ||
XM_008772017.1 | XP_008770239.1 | 4 - 5 | 1056 | 80 | ||
GenBank mRNA | D38629.1 | - | 4 - 5 | 577 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |