Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012599.2 | NP_036731.2 | 4 - 5 | 419 | 91 | |
GenBank mRNA | BC088159.1 | - | 4 - 5 | 419 | 7326244 | 91 |
FQ218582.1 | - | 4 - 5 | 447 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |