Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012600.2 | NP_036732.2 | 5 - 6 | 597 | 82 | |
XM_008766439.1 | XP_008764661.1 | 5 - 6 | 894 | 82 | ||
GenBank mRNA | CO560778.1 | - | 1 - 2 | 94 | 82 | |
FQ228526.1 | - | 5 - 6 | 628 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |