Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012780.1 | NP_036912.1 | 12 - 13 | 1178 | 112 | |
XM_006232850.2 | XP_006232912.1 | 12 - 13 | 1305 | 112 | ||
XM_006232852.2 | XP_006232914.1 | 12 - 13 | 1395 | 112 | ||
XM_006232853.2 | XP_006232915.1 | 11 - 12 | 1260 | 112 | ||
XM_008761280.1 | XP_008759502.1 | 12 - 13 | 1305 | 112 | ||
XM_008761281.1 | XP_008759503.1 | 11 - 12 | 1260 | 112 | ||
GenBank mRNA | AY264361.1 | - | 12 - 13 | 1178 | 112 | |
AY264362.1 | - | 11 - 12 | 1133 | 112 | ||
AY264363.1 | - | 12 - 13 | 1037 | 112 | ||
AY264364.1 | - | 12 - 13 | 1163 | 112 | ||
AY264365.1 | - | 12 - 13 | 1178 | 112 | ||
U08986.1 | - | 7 - 8 | 885 | 112 | ||
U61184.1 | - | 12 - 13 | 1178 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |