Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083122.1 | NP_001076591.1 | 4 - 5 | 636 | 123 | |
NM_012890.2 | NP_037022.1 | 3 - 4 | 575 | 123 | ||
GenBank mRNA | BC061997.1 | - | 3 - 4 | 578 | 123 | |
BC168237.1 | - | 4 - 5 | 629 | 7938890 | 123 | |
U50927.1 | - | 3 - 4 | 575 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |