Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013077.1 | NP_037209.1 | 5 - 6 | 567 | 60 | |
XM_008759690.1 | XP_008757912.1 | 5 - 6 | 843 | 60 | ||
GenBank mRNA | AB011544.1 | - | 5 - 6 | 567 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |