Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017012.1 | NP_058708.1 | 7 - 8 | 1951 | 78 | |
XM_006229659.2 | XP_006229721.1 | 7 - 8 | 2278 | 78 | ||
GenBank mRNA | D10891.1 | - | 7 - 8 | 1951 | 78 | |
EU559292.1 | - | 6 - 7 | 1684 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |