Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017046.1 | NP_058742.1 | 9 - 10 | 1468 | 59 | |
GenBank mRNA | EU627517.1 | - | 8 - 9 | 1370 | 59 | |
U37539.1 | - | 9 - 10 | 1422 | 59 | ||
U37540.1 | - | 9 - 10 | 1422 | 59 | ||
X77933.1 | - | 9 - 10 | 1468 | 59 | ||
X78034.1 | - | 9 - 10 | 1451 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |