Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017065.1 | NP_058761.1 | 155 | 72 | ||
GenBank mRNA | X15468.1 | - | 155 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |