Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 4 - 5 | 567 | 73 | |
XM_008759108.1 | XP_008757330.1 | 5 - 6 | 2966 | 73 | ||
XM_008759109.1 | XP_008757331.1 | 4 - 5 | 554 | 73 | ||
XM_008759110.1 | XP_008757332.1 | 4 - 5 | 2831 | 73 | ||
XM_008759111.1 | XP_008757333.1 | 4 - 5 | 677 | 73 | ||
GenBank mRNA | D30041.1 | - | 4 - 5 | 567 | 73 | |
DQ198085.1 | - | 3 - 4 | 286 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Rattus norvegicus Inventoried Probe spans exons |