Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017103.2 | NP_058799.2 | 3 - 4 | 300 | 79 | |
GenBank mRNA | BC081815.1 | - | 3 - 4 | 300 | 7110428 | 79 |
L46816.1 | - | 315 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |