Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017166.1 | NP_058862.1 | 3 - 4 | 357 | 137 | |
XM_006239123.2 | XP_006239185.1 | 3 - 4 | 355 | 137 | ||
GenBank mRNA | BC062234.1 | - | 3 - 4 | 282 | 6917958 | 137 |
FQ211845.1 | - | 3 - 4 | 280 | 137 | ||
FQ212924.1 | - | 3 - 4 | 279 | 137 | ||
FQ213053.1 | - | 3 - 4 | 280 | 137 | ||
FQ214201.1 | - | 3 - 4 | 274 | 137 | ||
FQ214447.1 | - | 3 - 4 | 280 | 137 | ||
FQ220047.1 | - | 3 - 4 | 280 | 137 | ||
J04979.1 | - | 3 - 4 | 357 | 137 | ||
M27876.1 | - | 2 - 3 | 300 | 137 | ||
X94914.1 | - | 1 - 2 | 189 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |