Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017193.1 | NP_058889.1 | 9 - 10 | 1073 | 140 | |
XM_006253067.2 | XP_006253129.1 | 9 - 10 | 1161 | 140 | ||
GenBank mRNA | BC078864.1 | - | 9 - 10 | 1069 | 7109210 | 140 |
Z50144.1 | - | 9 - 10 | 1073 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |