Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017256.1 | NP_058952.1 | 13 - 14 | 2495 | 87 | |
GenBank mRNA | M77809.1 | - | 13 - 14 | 2495 | 87 | |
M80784.1 | - | 13 - 14 | 2480 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |