Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017302.2 | NP_058998.2 | 2 - 3 | 309 | 69 | |
XM_006241432.2 | XP_006241494.1 | 3 - 4 | 851 | 69 | ||
XM_006241433.2 | XP_006241495.1 | 4 - 5 | 2367 | 69 | ||
XM_006241434.2 | XP_006241496.1 | 4 - 5 | 718 | 69 | ||
XM_006241435.2 | XP_006241497.1 | 3 - 4 | 2331 | 69 | ||
XM_006241436.2 | XP_006241498.1 | 3 - 4 | 593 | 69 | ||
XM_006241437.2 | XP_006241499.1 | 3 - 4 | 2241 | 69 | ||
GenBank mRNA | BC081701.1 | - | 2 - 3 | 309 | 7130138 | 69 |
FQ218176.1 | - | 3 - 4 | 423 | 69 | ||
U62316.1 | - | 3 - 4 | 475 | 69 | ||
X97445.1 | - | 3 - 4 | 434 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |