Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017315.2 | NP_059011.1 | 5 - 6 | 495 | 60 | |
XM_006254602.2 | XP_006254664.1 | 7 - 8 | 876 | 60 | ||
GenBank mRNA | AF080452.1 | - | 5 - 6 | 495 | 60 | |
BC078851.1 | - | 5 - 6 | 495 | 7127602 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |