Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127339.1 | NP_001120811.1 | 7 - 8 | 1139 | 80 | |
NM_001127341.1 | NP_001120813.1 | 7 - 8 | 1139 | 80 | ||
NM_001127342.1 | NP_001120814.1 | 5 - 6 | 710 | 80 | ||
NM_001127343.1 | NP_001120815.1 | 5 - 6 | 710 | 80 | ||
NM_001127344.1 | NP_001120816.1 | 5 - 6 | 710 | 80 | ||
NM_019221.3 | NP_062094.1 | 7 - 8 | 1139 | 80 | ||
XM_006248500.2 | XP_006248562.1 | 7 - 8 | 1220 | 80 | ||
XM_008768791.1 | XP_008767013.1 | 8 - 9 | 1069 | 80 | ||
GenBank mRNA | AJ277446.1 | - | 6 - 7 | 941 | 80 | |
AJ277447.1 | - | 5 - 6 | 710 | 80 | ||
AJ277448.1 | - | 6 - 7 | 941 | 80 | ||
AJ277449.1 | - | 7 - 8 | 992 | 80 | ||
AJ277450.1 | - | 5 - 6 | 710 | 80 | ||
AJ277451.1 | - | 7 - 8 | 992 | 80 | ||
AJ277452.1 | - | 6 - 7 | 941 | 80 | ||
AJ277453.1 | - | 5 - 6 | 710 | 80 | ||
Y10258.2 | - | 7 - 8 | 1139 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |