Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019255.1 | NP_062128.1 | 2 - 3 | 316 | 88 | |
GenBank mRNA | FQ215004.1 | - | 2 - 3 | 413 | 88 | |
FQ215168.1 | - | 2 - 3 | 395 | 88 | ||
FQ215755.1 | - | 2 - 3 | 395 | 88 | ||
FQ216734.1 | - | 2 - 3 | 431 | 88 | ||
FQ224130.1 | - | 2 - 3 | 487 | 88 | ||
FQ224197.1 | - | 2 - 3 | 410 | 88 | ||
Y09453.1 | - | 2 - 3 | 316 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |