Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019255.1 | NP_062128.1 | 3 - 4 | 455 | 57 | |
GenBank mRNA | FQ215004.1 | - | 3 - 4 | 552 | 57 | |
FQ215168.1 | - | 3 - 4 | 534 | 57 | ||
FQ215755.1 | - | 3 - 4 | 534 | 57 | ||
FQ216734.1 | - | 3 - 4 | 570 | 57 | ||
FQ224197.1 | - | 3 - 4 | 549 | 57 | ||
Y09453.1 | - | 3 - 4 | 455 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |