Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019275.2 | NP_062148.1 | 8 - 9 | 1192 | 89 | |
GenBank mRNA | AB010954.1 | - | 8 - 9 | 1192 | 89 | |
AF056002.1 | - | 1264 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |