Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019375.1 | NP_062248.1 | 6 - 7 | 704 | 72 | |
GenBank mRNA | AF111179.1 | - | 6 - 7 | 704 | 72 | |
AF111180.1 | - | 4 - 5 | 1111 | 72 | ||
AF111181.1 | - | 6 - 7 | 703 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |