Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239476.2 | XP_006239538.1 | 1 - 2 | 480 | 84 | |
XM_006239479.2 | XP_006239541.1 | 1 - 2 | 480 | 84 | ||
XM_008764375.1 | XP_008762597.1 | 1 - 2 | 480 | 84 | ||
XM_008764376.1 | XP_008762598.1 | 1 - 2 | 480 | 84 | ||
XM_008764377.1 | XP_008762599.1 | 1 - 2 | 480 | 84 | ||
XM_008764378.1 | XP_008762600.1 | 1 - 2 | 480 | 84 | ||
XM_008764379.1 | XP_008762601.1 | 1 - 2 | 480 | 84 | ||
XM_008764380.1 | XP_008762602.1 | 1 - 2 | 480 | 84 | ||
GenBank mRNA | U46007.1 | - | 1 - 2 | 306 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |