Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134901.1 | NP_001128373.1 | 5 - 6 | 554 | 150 | |
NM_020072.1 | NP_064457.1 | 5 - 6 | 594 | 150 | ||
GenBank mRNA | DQ826426.1 | - | 5 - 6 | 554 | 150 | |
M32397.1 | - | 5 - 6 | 594 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |