Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022258.2 | NP_071594.2 | 3 - 4 | 412 | 83 | |
GenBank mRNA | AJ302031.1 | - | 3 - 4 | 412 | 83 | |
BC089771.1 | - | 3 - 4 | 395 | 7326280 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |