Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031557.2 | NP_113745.1 | 6 - 7 | 1107 | 95 | |
GenBank mRNA | BC061814.1 | - | 6 - 7 | 1107 | 5598666 | 95 |
U53855.1 | - | 6 - 7 | 1038 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |