Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 12 - 13 | 2016 | 73 | |
GenBank mRNA | AB248960.1 | - | 5 - 6 | 432 | 73 | |
AF083341.1 | - | 12 - 13 | 1452 | 73 | ||
AF135265.1 | - | 12 - 13 | 1870 | 73 | ||
AY330290.1 | - | 12 - 13 | 1341 | 73 | ||
AY330291.1 | - | 12 - 13 | 1341 | 73 | ||
AY330293.2 | - | 12 - 13 | 1341 | 73 | ||
AY330294.1 | - | 12 - 13 | 1341 | 73 | ||
AY344965.2 | - | 11 - 12 | 1753 | 73 | ||
U40603.1 | - | 12 - 13 | 2016 | 73 | ||
U55995.1 | - | 12 - 13 | 1494 | 73 | ||
U93052.1 | - | 12 - 13 | 1442 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |