Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031972.1 | NP_114178.1 | 6 - 7 | 976 | 64 | |
GenBank mRNA | BC070924.1 | - | 6 - 7 | 1009 | 7096634 | 64 |
J03637.1 | - | 6 - 7 | 976 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |