Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053896.2 | NP_446348.2 | 11 - 12 | 1414 | 65 | |
GenBank mRNA | BC098910.1 | - | 11 - 12 | 1406 | 7455646 | 65 |
U60063.1 | - | 11 - 12 | 1410 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |