Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190399.1 | NP_001177328.1 | 2 - 3 | 238 | 74 | |
NM_138915.1 | NP_620270.1 | 2 - 3 | 297 | 74 | ||
GenBank mRNA | AF303658.1 | - | 2 - 3 | 297 | 74 | |
FQ213422.1 | - | 2 - 3 | 212 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |