Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170787.2 | NP_740768.1 | 3 - 4 | 1099 | 70 | |
GenBank mRNA | AF541925.1 | - | 3 - 4 | 1099 | 70 | |
AY049038.1 | - | 2 - 3 | 1133 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |