Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022618.1 | NP_072140.1 | 11 - 12 | 3496 | 56 | |
GenBank mRNA | AF139518.1 | - | 11 - 12 | 3496 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |