Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053804.2 | NP_446256.2 | 5 - 6 | 807 | 64 | |
XM_006230634.2 | XP_006230696.1 | 6 - 7 | 2105 | 64 | ||
XM_008760054.1 | XP_008758276.1 | 6 - 7 | 1848 | 64 | ||
XM_008760055.1 | XP_008758277.1 | 6 - 7 | 1719 | 64 | ||
XM_008760056.1 | XP_008758278.1 | 6 - 7 | 925 | 64 | ||
GenBank mRNA | AF259502.1 | - | 1 - 2 | 173 | 64 | |
AF302842.2 | - | 5 - 6 | 807 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons |