Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080903.1 | NP_543179.1 | 4 - 5 | 685 | 91 | |
GenBank mRNA | AY059627.1 | - | 4 - 5 | 685 | 91 | |
BC061824.1 | - | 4 - 5 | 672 | 5599072 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |