Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024384.2 | NP_077360.2 | 10 - 11 | 943 | 87 | |
GenBank mRNA | AF275266.1 | - | 3 - 4 | 208 | 87 | |
BC079149.1 | - | 10 - 11 | 943 | 7128541 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |