Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181084.2 | NP_851598.1 | 2 - 3 | 586 | 66 | |
XM_006237868.2 | XP_006237930.1 | 3 - 4 | 717 | 66 | ||
GenBank mRNA | AB107917.1 | - | 2 - 3 | 586 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |