Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181477.2 | NP_852142.1 | 2 - 3 | 152 | 68 | |
XM_006245872.2 | XP_006245934.1 | 2 - 3 | 570 | 68 | ||
GenBank mRNA | AB074516.1 | - | 2 - 3 | 152 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |