Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022407.3 | NP_071852.2 | 5 - 6 | 533 | 121 | |
GenBank mRNA | AF001896.1 | - | 5 - 6 | 533 | 121 | |
AF001897.1 | - | 5 - 6 | 533 | 121 | ||
AF001898.1 | - | 5 - 6 | 533 | 121 | ||
BC061526.1 | - | 5 - 6 | 552 | 6921926 | 121 | |
U79118.1 | - | 5 - 6 | 532 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |