Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183055.1 | NP_898878.1 | 1 - 2 | 224 | 94 | |
GenBank mRNA | AY161005.1 | - | 1 - 2 | 224 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |