Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183329.3 | NP_899158.3 | 3 - 4 | 411 | 58 | |
GenBank mRNA | AY026354.1 | - | 3 - 4 | 359 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |