Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250164.2 | XP_006250226.1 | 16 - 17 | 2025 | 115 | |
XM_006250166.2 | XP_006250228.1 | 15 - 16 | 2004 | 115 | ||
XM_008769657.1 | XP_008767879.1 | 16 - 17 | 2037 | 115 | ||
XM_008769658.1 | XP_008767880.1 | 15 - 16 | 2016 | 115 | ||
XM_008769659.1 | XP_008767881.1 | 16 - 17 | 2037 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |