Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031643.4 | NP_113831.1 | 6 - 7 | 745 | 67 | |
GenBank mRNA | BC089772.1 | - | 6 - 7 | 739 | 7382030 | 67 |
D13341.1 | - | 6 - 7 | 728 | 67 | ||
D14591.1 | - | 6 - 7 | 698 | 67 | ||
X62313.1 | - | 6 - 7 | 735 | 67 | ||
Z16415.1 | - | 6 - 7 | 728 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |