Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012899.2 | NP_037031.1 | 2 - 3 | 275 | 64 | |
XM_006238234.2 | XP_006238296.1 | 2 - 3 | 868 | 64 | ||
XM_006238235.1 | XP_006238297.1 | 2 - 3 | 365 | 64 | ||
GenBank mRNA | BC061806.1 | - | 2 - 3 | 275 | 5598340 | 64 |
FQ210250.1 | - | 2 - 3 | 220 | 64 | ||
FQ218546.1 | - | 2 - 3 | 220 | 64 | ||
FQ219571.1 | - | 2 - 3 | 220 | 64 | ||
FQ231154.1 | - | 2 - 3 | 251 | 64 | ||
FQ232546.1 | - | 2 - 3 | 251 | 64 | ||
FQ233724.1 | - | 2 - 3 | 301 | 64 | ||
FQ234245.1 | - | 2 - 3 | 251 | 64 | ||
FQ234573.1 | - | 2 - 3 | 299 | 64 | ||
X04959.1 | - | 1 - 2 | 157 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |