Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017138.2 | NP_058834.1 | 3 - 4 | 316 | 57 | |
XM_008766672.1 | XP_008764894.1 | 3 - 4 | 389 | 57 | ||
GenBank mRNA | BC060578.1 | - | 3 - 4 | 334 | 6919235 | 57 |
D25224.1 | - | 3 - 4 | 317 | 57 | ||
FJ386454.1 | - | 2 - 3 | 297 | 57 | ||
FQ209463.1 | - | 3 - 4 | 330 | 57 | ||
FQ209469.1 | - | 3 - 4 | 330 | 57 | ||
FQ209490.1 | - | 3 - 4 | 331 | 57 | ||
FQ209667.1 | - | 3 - 4 | 330 | 57 | ||
FQ209729.1 | - | 3 - 4 | 330 | 57 | ||
FQ212660.1 | - | 3 - 4 | 330 | 57 | ||
FQ212893.1 | - | 3 - 4 | 330 | 57 | ||
FQ213275.1 | - | 3 - 4 | 330 | 57 | ||
FQ213317.1 | - | 3 - 4 | 330 | 57 | ||
FQ213334.1 | - | 3 - 4 | 330 | 57 | ||
FQ213623.1 | - | 3 - 4 | 333 | 57 | ||
FQ213835.1 | - | 3 - 4 | 331 | 57 | ||
FQ213894.1 | - | 3 - 4 | 330 | 57 | ||
FQ215242.1 | - | 3 - 4 | 330 | 57 | ||
FQ215292.1 | - | 3 - 4 | 331 | 57 | ||
FQ216085.1 | - | 3 - 4 | 330 | 57 | ||
FQ216130.1 | - | 3 - 4 | 330 | 57 | ||
FQ216245.1 | - | 3 - 4 | 330 | 57 | ||
FQ216788.1 | - | 3 - 4 | 332 | 57 | ||
FQ216997.1 | - | 3 - 4 | 330 | 57 | ||
FQ217664.1 | - | 3 - 4 | 330 | 57 | ||
FQ219586.1 | - | 3 - 4 | 330 | 57 | ||
FQ219785.1 | - | 3 - 4 | 333 | 57 | ||
FQ220012.1 | - | 3 - 4 | 330 | 57 | ||
FQ220017.1 | - | 3 - 4 | 328 | 57 | ||
FQ220075.1 | - | 3 - 4 | 331 | 57 | ||
FQ220084.1 | - | 3 - 4 | 333 | 57 | ||
FQ220126.1 | - | 3 - 4 | 330 | 57 | ||
FQ220134.1 | - | 3 - 4 | 330 | 57 | ||
FQ220171.1 | - | 3 - 4 | 329 | 57 | ||
FQ220184.1 | - | 3 - 4 | 330 | 57 | ||
FQ220347.1 | - | 3 - 4 | 332 | 57 | ||
FQ220461.1 | - | 3 - 4 | 311 | 57 | ||
FQ220468.1 | - | 3 - 4 | 330 | 57 | ||
FQ220661.1 | - | 3 - 4 | 333 | 57 | ||
FQ220822.1 | - | 3 - 4 | 330 | 57 | ||
FQ220904.1 | - | 3 - 4 | 331 | 57 | ||
FQ221103.1 | - | 3 - 4 | 330 | 57 | ||
FQ221443.1 | - | 3 - 4 | 331 | 57 | ||
FQ222093.1 | - | 3 - 4 | 332 | 57 | ||
FQ222280.1 | - | 3 - 4 | 330 | 57 | ||
FQ222542.1 | - | 3 - 4 | 332 | 57 | ||
FQ222757.1 | - | 3 - 4 | 328 | 57 | ||
FQ224747.1 | - | 3 - 4 | 330 | 57 | ||
FQ225510.1 | - | 3 - 4 | 331 | 57 | ||
FQ226054.1 | - | 3 - 4 | 330 | 57 | ||
FQ226408.1 | - | 3 - 4 | 330 | 57 | ||
FQ227056.1 | - | 3 - 4 | 330 | 57 | ||
FQ227293.1 | - | 3 - 4 | 330 | 57 | ||
FQ227600.1 | - | 3 - 4 | 331 | 57 | ||
FQ228002.1 | - | 3 - 4 | 330 | 57 | ||
FQ228212.1 | - | 3 - 4 | 331 | 57 | ||
FQ229290.1 | - | 3 - 4 | 332 | 57 | ||
FQ229305.1 | - | 3 - 4 | 332 | 57 | ||
FQ229367.1 | - | 3 - 4 | 331 | 57 | ||
FQ229516.1 | - | 3 - 4 | 330 | 57 | ||
FQ229579.1 | - | 3 - 4 | 330 | 57 | ||
FQ229598.1 | - | 3 - 4 | 331 | 57 | ||
FQ229712.1 | - | 3 - 4 | 330 | 57 | ||
FQ229787.1 | - | 3 - 4 | 330 | 57 | ||
FQ229859.1 | - | 3 - 4 | 328 | 57 | ||
FQ229890.1 | - | 3 - 4 | 330 | 57 | ||
FQ229945.1 | - | 3 - 4 | 330 | 57 | ||
FQ229970.1 | - | 3 - 4 | 330 | 57 | ||
FQ230088.1 | - | 3 - 4 | 57 | |||
FQ230106.1 | - | 3 - 4 | 330 | 57 | ||
FQ230180.1 | - | 3 - 4 | 331 | 57 | ||
FQ230258.1 | - | 3 - 4 | 329 | 57 | ||
FQ231469.1 | - | 2 - 3 | 147 | 57 | ||
FQ231622.1 | - | 3 - 4 | 328 | 57 | ||
FQ232568.1 | - | 3 - 4 | 331 | 57 | ||
FQ233068.1 | - | 3 - 4 | 330 | 57 | ||
FQ233112.1 | - | 3 - 4 | 330 | 57 | ||
FQ233340.1 | - | 3 - 4 | 332 | 57 | ||
FQ233547.1 | - | 3 - 4 | 331 | 57 | ||
FQ233919.1 | - | 3 - 4 | 330 | 57 | ||
FQ234511.1 | - | 3 - 4 | 330 | 57 | ||
FQ234742.1 | - | 3 - 4 | 330 | 57 | ||
FQ234765.1 | - | 3 - 4 | 330 | 57 | ||
FQ234772.1 | - | 3 - 4 | 330 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |