Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053713.1 | NP_446165.1 | 3 - 4 | 1183 | 119 | |
GenBank mRNA | AF390546.1 | - | 3 - 4 | 1183 | 119 | |
BC085720.1 | - | 3 - 4 | 1576 | 7190219 | 119 | |
FQ233252.1 | - | 1 - 2 | 550 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |