Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053748.1 | NP_446200.1 | 15 - 16 | 1890 | 66 | |
XM_006230633.2 | XP_006230695.1 | 16 - 17 | 2077 | 66 | ||
GenBank mRNA | BC107673.1 | - | 16 - 17 | 1901 | 7938129 | 66 |
D89340.2 | - | 15 - 16 | 1890 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |