Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053867.1 | NP_446319.1 | 5 - 6 | 581 | 103 | |
GenBank mRNA | BC086358.1 | - | 5 - 6 | 592 | 7303675 | 103 |
FQ210845.1 | - | 4 - 5 | 495 | 103 | ||
FQ217312.1 | - | 5 - 6 | 609 | 103 | ||
FQ217485.1 | - | 5 - 6 | 609 | 103 | ||
FQ220662.1 | - | 5 - 6 | 609 | 103 | ||
FQ220913.1 | - | 5 - 6 | 612 | 103 | ||
FQ221184.1 | - | 5 - 6 | 609 | 103 | ||
FQ221191.1 | - | 5 - 6 | 598 | 103 | ||
FQ221220.1 | - | 5 - 6 | 609 | 103 | ||
FQ221227.1 | - | 5 - 6 | 609 | 103 | ||
FQ221357.1 | - | 5 - 6 | 609 | 103 | ||
FQ221503.1 | - | 5 - 6 | 609 | 103 | ||
FQ221524.1 | - | 5 - 6 | 609 | 103 | ||
FQ221695.1 | - | 5 - 6 | 609 | 103 | ||
FQ221696.1 | - | 5 - 6 | 610 | 103 | ||
FQ221929.1 | - | 2 - 3 | 227 | 103 | ||
FQ221991.1 | - | 3 - 4 | 250 | 103 | ||
FQ222296.1 | - | 5 - 6 | 609 | 103 | ||
FQ222704.1 | - | 5 - 6 | 608 | 103 | ||
FQ222827.1 | - | 5 - 6 | 609 | 103 | ||
FQ223078.1 | - | 5 - 6 | 609 | 103 | ||
FQ223172.1 | - | 5 - 6 | 610 | 103 | ||
FQ223294.1 | - | 5 - 6 | 616 | 103 | ||
FQ223538.1 | - | 5 - 6 | 609 | 103 | ||
FQ223612.1 | - | 5 - 6 | 598 | 103 | ||
FQ224095.1 | - | 5 - 6 | 609 | 103 | ||
FQ224271.1 | - | 5 - 6 | 610 | 103 | ||
FQ224320.1 | - | 5 - 6 | 608 | 103 | ||
FQ224622.1 | - | 5 - 6 | 545 | 103 | ||
FQ224742.1 | - | 5 - 6 | 609 | 103 | ||
FQ226252.1 | - | 5 - 6 | 609 | 103 | ||
FQ227715.1 | - | 5 - 6 | 610 | 103 | ||
FQ227806.1 | - | 5 - 6 | 638 | 103 | ||
FQ228514.1 | - | 5 - 6 | 609 | 103 | ||
FQ228556.1 | - | 5 - 6 | 609 | 103 | ||
FQ228624.1 | - | 5 - 6 | 608 | 103 | ||
FQ228630.1 | - | 5 - 6 | 609 | 103 | ||
FQ228644.1 | - | 5 - 6 | 606 | 103 | ||
FQ229093.1 | - | 5 - 6 | 609 | 103 | ||
FQ229217.1 | - | 5 - 6 | 609 | 103 | ||
FQ229279.1 | - | 5 - 6 | 609 | 103 | ||
FQ229346.1 | - | 5 - 6 | 609 | 103 | ||
FQ229353.1 | - | 5 - 6 | 609 | 103 | ||
FQ230785.1 | - | 5 - 6 | 609 | 103 | ||
FQ231339.1 | - | 5 - 6 | 607 | 103 | ||
FQ231778.1 | - | 5 - 6 | 609 | 103 | ||
FQ232768.1 | - | 5 - 6 | 610 | 103 | ||
FQ233320.1 | - | 5 - 6 | 640 | 103 | ||
FQ233909.1 | - | 5 - 6 | 612 | 103 | ||
FQ234047.1 | - | 5 - 6 | 599 | 103 | ||
FQ234298.1 | - | 5 - 6 | 609 | 103 | ||
FQ234990.1 | - | 5 - 6 | 609 | 103 | ||
FQ235307.1 | - | 5 - 6 | 609 | 103 | ||
U20525.1 | - | 5 - 6 | 581 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |