Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145878.1 | NP_665885.1 | 1 - 2 | 118 | 90 | |
GenBank mRNA | FQ210725.1 | - | 1 - 2 | 128 | 90 | |
S69874.1 | - | 1 - 2 | 120 | 90 | ||
S83247.1 | - | 1 - 2 | 135 | 90 | ||
U13253.1 | - | 1 - 2 | 118 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |