Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007740.1 | NP_001007741.1 | 3 - 4 | 307 | 119 | |
XM_006256460.2 | XP_006256522.1 | 3 - 4 | 430 | 119 | ||
XM_008772892.1 | XP_008771114.1 | 3 - 4 | 435 | 119 | ||
GenBank mRNA | BC079150.1 | - | 3 - 4 | 307 | 7128580 | 119 |
FQ211877.1 | - | 3 - 4 | 320 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |