Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 6 - 7 | 820 | 73 | |
XM_006256359.2 | XP_006256421.1 | 6 - 7 | 871 | 73 | ||
XM_006256360.2 | XP_006256422.1 | 5 - 6 | 667 | 73 | ||
XM_006256361.2 | XP_006256423.1 | 6 - 7 | 894 | 73 | ||
XM_006256362.2 | XP_006256424.1 | 6 - 7 | 786 | 73 | ||
XM_006256363.1 | XP_006256425.1 | 2 - 3 | 228 | 73 | ||
XM_006256364.1 | XP_006256426.1 | 6 - 7 | 780 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |